Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5B2
Entrez Gene ID
90113
Gene Name
von Willebrand factor A domain containing 5B2
Gene Aliases
-
Chromosome Location
Chr.3: 184229593 - 184242329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320373.1 NP_001307302.1 11 - 12 1713 111
NM_138345.2 NP_612354.1 13 - 14 2301 111
XM_011513288.2 XP_011511590.1 14 - 15 2940 111
XM_011513289.2 XP_011511591.1 14 - 15 3023 111
XM_011513290.2 XP_011511592.1 14 - 15 2486 111
XM_011513291.2 XP_011511593.1 13 - 14 3149 111
XM_011513292.2 XP_011511594.1 13 - 14 3140 111
GenBank mRNA AK293885.1 - 11 - 12 1713 111
AL834499.1 - 7 - 8 1133 111
BC001200.1 - 8 - 9 1480 111
BC136609.1 - 13 - 14 2301 111

View Full Product Details