The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320373.1 | NP_001307302.1 | 6 - 7 | 725 | 55 |
NM_138345.2 | NP_612354.1 | 8 - 9 | 1316 | 55 | |
XM_011513288.2 | XP_011511590.1 | 9 - 10 | 1952 | 55 | |
XM_011513289.2 | XP_011511591.1 | 9 - 10 | 2035 | 55 | |
XM_011513290.2 | XP_011511592.1 | 9 - 10 | 1498 | 55 | |
XM_011513291.2 | XP_011511593.1 | 8 - 9 | 2164 | 55 | |
XM_011513292.2 | XP_011511594.1 | 8 - 9 | 2152 | 55 | |
GenBank mRNA | AK293885.1 | - | 6 - 7 | 725 | 55 |
AL834499.1 | - | 2 - 3 | 148 | 55 | |
BC001200.1 | - | 3 - 4 | 495 | 55 | |
BC136609.1 | - | 8 - 9 | 1316 | 55 | |
DC310771.1 | - | 5 - 6 | 568 | 55 |