Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
Chromosome Location
Chr.3: 132558138 - 132660131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032169.4 NP_115545.3 12 - 13 1906 72
NR_132426.1 12 - 13 1906 72
NR_132427.1 12 - 13 1906 72
NR_132428.1 10 - 11 1689 72
GenBank mRNA AJ608287.1 - 12 - 13 1857 72
AK022654.1 - 5 - 6 500 72
AK131265.1 - 14 - 15 1703 72
AK298627.1 - 4 - 5 713 72
AL832873.1 - 12 - 13 1857 72
AL833721.1 - 12 - 13 1424 72
AX803783.1 - 12 - 13 1763 72
AX803785.1 - 12 - 13 1526 72
BC019607.1 - 7 - 8 802 72
BC125204.1 - 12 - 13 1841 72
BC125205.1 - 10 - 11 1624 72

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
Chromosome Location
Chr.3: 132558138 - 132722459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037804.1 38 - 39 5528 72
GenBank mRNA AB082531.1 - 38 - 39 5528 72

View Full Product Details