The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005247711.4 | XP_005247768.1 | 8 - 9 | 959 | 84 |
XM_006713733.3 | XP_006713796.1 | 8 - 9 | 961 | 84 | |
XM_017007059.1 | XP_016862548.1 | 4 - 5 | 1010 | 84 | |
GenBank mRNA | AK027607.1 | - | 4 - 5 | 410 | 84 |