The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002612.3 | NP_002603.1 | 4 - 5 | 848 | 71 |
GenBank mRNA | AK096428.1 | - | 5 - 6 | 647 | 71 |
AK223328.1 | - | 4 - 5 | 744 | 71 | |
AK312424.1 | - | 4 - 5 | 754 | 71 | |
BC040239.1 | - | 4 - 5 | 728 | 71 | |
U54617.1 | - | 4 - 5 | 750 | 71 |