The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142326.1 | NP_001135798.1 | 9 - 10 | 1027 | 87 |
NM_001142327.1 | NP_001135799.1 | 10 - 11 | 1144 | 87 | |
NM_021145.3 | NP_066968.3 | 12 - 13 | 1381 | 87 | |
NR_024549.1 | 10 - 11 | 1355 | 87 | ||
NR_024550.1 | 10 - 11 | 1316 | 87 | ||
XM_011516735.2 | XP_011515037.1 | 10 - 11 | 2066 | 87 | |
XM_011516737.1 | XP_011515039.1 | 11 - 12 | 1191 | 87 | |
XM_011516739.2 | XP_011515041.1 | 9 - 10 | 956 | 87 | |
XM_017012865.1 | XP_016868354.1 | 11 - 12 | 2160 | 87 | |
XM_017012866.1 | XP_016868355.1 | 9 - 10 | 1943 | 87 | |
XM_017012867.1 | XP_016868356.1 | 8 - 9 | 2611 | 87 | |
XM_017012868.1 | XP_016868357.1 | 11 - 12 | 1191 | 87 | |
XM_017012869.1 | XP_016868358.1 | 12 - 13 | 1220 | 87 | |
XM_017012870.1 | XP_016868359.1 | 9 - 10 | 959 | 87 | |
XM_017012871.1 | XP_016868360.1 | 11 - 12 | 2332 | 87 | |
XM_017012872.1 | XP_016868361.1 | 10 - 11 | 2238 | 87 | |
XM_017012873.1 | XP_016868362.1 | 11 - 12 | 2127 | 87 | |
XM_017012874.1 | XP_016868363.1 | 11 - 12 | 1363 | 87 | |
XM_017012875.1 | XP_016868364.1 | 11 - 12 | 2166 | 87 | |
XM_017012876.1 | XP_016868365.1 | 9 - 10 | 1106 | 87 | |
XM_017012877.1 | XP_016868366.1 | 10 - 11 | 1225 | 87 | |
XM_017012878.1 | XP_016868367.1 | 12 - 13 | 2260 | 87 | |
GenBank mRNA | AB209230.1 | - | 9 - 10 | 1150 | 87 |
AF084530.1 | - | 10 - 11 | 1093 | 87 | |
AF202144.1 | - | 10 - 11 | 1265 | 87 | |
AF202145.1 | - | 10 - 11 | 1304 | 87 | |
AK126664.1 | - | 10 - 11 | 1811 | 87 | |
AK295144.1 | - | 9 - 10 | 1167 | 87 | |
AK296211.1 | - | 9 - 10 | 920 | 87 | |
AK314622.1 | - | 10 - 11 | 1080 | 87 | |
BC007418.2 | - | 1 - 2 | 93 | 87 | |
BC007447.2 | - | 1 - 2 | 93 | 87 | |
BC070064.1 | - | 12 - 13 | 1327 | 87 |