Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DMTF1
- Entrez Gene ID
-
9988
- Gene Name
- cyclin D binding myb like transcription factor 1
- Gene Aliases
-
DMP1, DMTF, MRUL, hDMP1
- Chromosome Location
-
Chr.7: 87152361 - 87196332 on Build GRCh38
View Full Product Details