The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102469.1 | NP_001095939.1 | 4 - 5 | 532 | 127 |
XM_005270049.3 | XP_005270106.1 | 6 - 7 | 824 | 127 | |
XM_011540083.2 | XP_011538385.1 | 6 - 7 | 861 | 127 | |
XM_011540084.2 | XP_011538386.1 | 5 - 6 | 746 | 127 | |
XM_011540085.2 | XP_011538387.1 | 6 - 7 | 861 | 127 | |
XM_011540086.2 | XP_011538388.1 | 6 - 7 | 861 | 127 | |
XM_017016549.1 | XP_016872038.1 | 6 - 7 | 861 | 127 | |
GenBank mRNA | EF426483.1 | - | 4 - 5 | 532 | 127 |