The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004841.3 | NP_004832.1 | 5 - 6 | 612 | 84 |
NM_170692.2 | NP_733793.2 | 7 - 8 | 1695 | 84 | |
XM_005245622.3 | XP_005245679.1 | 5 - 6 | 1268 | 84 | |
XM_011510166.2 | XP_011508468.1 | 7 - 8 | 1695 | 84 | |
XM_011510167.2 | XP_011508469.1 | 7 - 8 | 1695 | 84 | |
XM_017002849.1 | XP_016858338.1 | 7 - 8 | 4301 | 84 | |
XM_017002850.1 | XP_016858339.1 | 7 - 8 | 4300 | 84 | |
XM_017002851.1 | XP_016858340.1 | 7 - 8 | 4300 | 84 | |
XM_017002852.1 | XP_016858341.1 | 7 - 8 | 4299 | 84 | |
XM_017002853.1 | XP_016858342.1 | 7 - 8 | 1739 | 84 | |
XM_017002854.1 | XP_016858343.1 | 5 - 6 | 1271 | 84 | |
XM_017002855.1 | XP_016858344.1 | 5 - 6 | 1270 | 84 | |
GenBank mRNA | AB209789.1 | - | 7 - 8 | 1603 | 84 |
AF047711.1 | - | 5 - 6 | 612 | 84 | |
AK075169.1 | - | 7 - 8 | 1695 | 84 | |
AK290308.1 | - | 5 - 6 | 555 | 84 | |
BC041045.1 | - | - | 1248 | 84 | |
BC063442.1 | - | 7 - 8 | 1255 | 84 | |
BC078167.1 | - | 7 - 8 | 1317 | 84 | |
BC110611.2 | - | 7 - 8 | 927 | 84 |