Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
Chromosome Location
Chr.16: 50317563 - 50368969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173984.2 NP_001167455.1 16 - 17 2068 73
NM_013263.4 NP_037395.2 16 - 17 2065 73
XM_011523046.2 XP_011521348.1 16 - 17 2154 73
XM_011523047.2 XP_011521349.1 16 - 17 2151 73
XM_011523048.2 XP_011521350.1 15 - 16 2046 73
XM_011523049.2 XP_011521351.1 15 - 16 2043 73
XM_011523050.2 XP_011521352.1 12 - 13 2174 73
XM_017023179.1 XP_016878668.1 15 - 16 1860 73
XM_017023180.1 XP_016878669.1 15 - 16 1857 73
XM_017023181.1 XP_016878670.1 12 - 13 2188 73
GenBank mRNA AF152604.1 - 16 - 17 1958 73
AF213969.1 - 16 - 17 1913 73
AJ271881.1 - 16 - 17 2023 73
AK027308.1 - 9 - 10 1036 73
AK074613.1 - 10 - 11 1202 73
BC001611.1 - 12 - 13 1329 73
BC050728.1 - 17 - 18 1972 73
BC094706.1 - 17 - 18 1942 73
CD677718.1 - 1 - 2 92 73

View Full Product Details