The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167749.2 | NP_001161221.1 | 16 - 17 | 2283 | 107 |
NM_001297772.1 | NP_001284701.1 | 19 - 20 | 2775 | 107 | |
NM_018417.5 | NP_060887.2 | 19 - 20 | 2738 | 107 | |
XM_005245330.4 | XP_005245387.1 | 19 - 20 | 4068 | 107 | |
XM_006711449.3 | XP_006711512.1 | 19 - 20 | 4068 | 107 | |
XM_011509760.2 | XP_011508062.1 | 20 - 21 | 4208 | 107 | |
XM_011509762.2 | XP_011508064.1 | 18 - 19 | 3924 | 107 | |
XM_011509763.2 | XP_011508065.1 | 19 - 20 | 4068 | 107 | |
XM_011509766.2 | XP_011508068.1 | 19 - 20 | 4068 | 107 | |
XM_017001778.1 | XP_016857267.1 | 19 - 20 | 4068 | 107 | |
GenBank mRNA | AF176813.2 | - | 19 - 20 | 2614 | 107 |
AF271058.1 | - | 18 - 19 | 2429 | 107 | |
AF299350.1 | - | 19 - 20 | 2614 | 107 | |
AF331033.1 | - | 19 - 20 | 2651 | 107 | |
AK302884.1 | - | 16 - 17 | 2272 | 107 | |
AK310395.1 | - | 19 - 20 | 2625 | 107 | |
AL035122.1 | - | 1 - 2 | 135 | 107 | |
BC117366.1 | - | 19 - 20 | 2518 | 107 | |
BC117372.1 | - | 19 - 20 | 2518 | 107 | |
GQ472219.1 | - | 19 - 20 | 2518 | 107 |