The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101669.1 | NP_001095139.1 | 13 - 14 | 1461 | 137 |
NM_003866.2 | NP_003857.2 | 14 - 15 | 1560 | 137 | |
XM_005263323.3 | XP_005263380.1 | 6 - 7 | 669 | 137 | |
XM_005263325.3 | XP_005263382.1 | 6 - 7 | 642 | 137 | |
XM_011532391.2 | XP_011530693.1 | 13 - 14 | 1545 | 137 | |
XM_011532392.2 | XP_011530694.1 | 13 - 14 | 1311 | 137 | |
XM_017008794.1 | XP_016864283.1 | 13 - 14 | 1545 | 137 | |
XM_017008795.1 | XP_016864284.1 | 13 - 14 | 1545 | 137 | |
XM_017008796.1 | XP_016864285.1 | 13 - 14 | 1545 | 137 | |
XM_017008797.1 | XP_016864286.1 | 6 - 7 | 665 | 137 | |
XM_017008798.1 | XP_016864287.1 | 6 - 7 | 642 | 137 | |
GenBank mRNA | AK298118.1 | - | 6 - 7 | 610 | 137 |
AK300891.1 | - | 6 - 7 | 675 | 137 | |
AK302936.1 | - | 12 - 13 | 1227 | 137 | |
BC005274.1 | - | 13 - 14 | 1236 | 137 | |
BC072447.1 | - | 12 - 13 | 1368 | 137 | |
BC110918.1 | - | 12 - 13 | 1318 | 137 | |
BC133005.1 | - | 10 - 11 | 1016 | 137 | |
BC144255.1 | - | 10 - 11 | 1016 | 137 | |
BX649090.1 | - | 14 - 15 | 1401 | 137 | |
U96922.1 | - | 10 - 11 | 137 |