Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OCIAD2
Entrez Gene ID
132299
Gene Name
OCIA domain containing 2
Gene Aliases
-
Chromosome Location
Chr.4: 48885380 - 48906828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014446.2 NP_001014446.1 6 - 7 608 146
NM_001286774.1 NP_001273703.1 5 - 6 511 146
GenBank mRNA AK298305.1 - 6 - 7 577 146
AY472117.1 - 6 - 7 610 146
BI546814.1 - 6 - 7 613 146
BU508058.1 - 5 - 6 399 146
CR749310.1 - 5 - 6 1669 146

View Full Product Details