The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002039.3 | NP_002030.2 | 4 - 5 | 1556 | 103 |
NM_207123.2 | NP_997006.1 | 4 - 5 | 1556 | 103 | |
XM_006714167.3 | XP_006714230.1 | 5 - 6 | 2454 | 103 | |
XM_006714168.3 | XP_006714231.1 | 5 - 6 | 2118 | 103 | |
XM_017007966.1 | XP_016863455.1 | 6 - 7 | 8281 | 103 | |
XM_017007967.1 | XP_016863456.1 | 5 - 6 | 1366 | 103 | |
XM_017007968.1 | XP_016863457.1 | 5 - 6 | 8198 | 103 | |
XM_017007969.1 | XP_016863458.1 | 5 - 6 | 3530 | 103 | |
XM_017007970.1 | XP_016863459.1 | 5 - 6 | 3285 | 103 | |
XM_017007971.1 | XP_016863460.1 | 4 - 5 | 3204 | 103 | |
GenBank mRNA | AK289767.1 | - | 4 - 5 | 1499 | 103 |
AK295684.1 | - | 4 - 5 | 1200 | 103 | |
AK314635.1 | - | 4 - 5 | 1236 | 103 | |
BC064848.1 | - | 4 - 5 | 1420 | 103 | |
U43885.1 | - | 4 - 5 | 1318 | 103 |