The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317769.1 | NP_001304698.1 | 2 - 3 | 115 | |
NM_012108.3 | NP_036240.1 | 2 - 3 | 115 | ||
XM_017008018.1 | XP_016863507.1 | 2 - 3 | 115 | ||
GenBank mRNA | AB023483.1 | - | 2 - 3 | 115 | |
AK313676.1 | - | 2 - 3 | 115 | ||
BC014958.1 | - | 2 - 3 | 115 | ||
BX372307.2 | - | 4 - 5 | 372 | 115 |