The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317769.1 | NP_001304698.1 | 6 - 7 | 763 | 95 |
NM_012108.3 | NP_036240.1 | 6 - 7 | 763 | 95 | |
GenBank mRNA | AB023483.1 | - | 6 - 7 | 733 | 95 |
AK313676.1 | - | 6 - 7 | 712 | 95 | |
BC014958.1 | - | 6 - 7 | 682 | 95 |