Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY3
Entrez Gene ID
23001
Gene Name
WD repeat and FYVE domain containing 3
Gene Aliases
ALFY, ZFYVE25
Chromosome Location
Chr.4: 84669537 - 84966391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014991.4 NP_055806.2 36 - 37 6378 110
XM_005262858.4 XP_005262915.1 38 - 39 6419 110
XM_011531757.2 XP_011530059.1 37 - 38 6330 110
XM_011531759.2 XP_011530061.1 36 - 37 6292 110
XM_011531760.2 XP_011530062.1 36 - 37 6230 110
XM_011531761.2 XP_011530063.1 37 - 38 6285 110
XM_011531762.2 XP_011530064.1 37 - 38 6330 110
XM_011531763.2 XP_011530065.1 35 - 36 6176 110
XM_011531764.2 XP_011530066.1 36 - 37 6246 110
XM_011531765.2 XP_011530067.1 34 - 35 6032 110
XM_011531766.2 XP_011530068.1 37 - 38 6330 110
XM_011531767.2 XP_011530069.1 37 - 38 6330 110
XM_017007906.1 XP_016863395.1 39 - 40 6547 110
XM_017007907.1 XP_016863396.1 39 - 40 6544 110
XM_017007908.1 XP_016863397.1 38 - 39 6476 110
XM_017007909.1 XP_016863398.1 24 - 25 4172 110
GenBank mRNA AB023210.2 - 1 - 2 61 110
AF538685.1 - 35 - 36 6270 110
BC150246.1 - 1 - 2 82 110

View Full Product Details