The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 45 - 46 | 7683 | 92 |
XM_005262858.4 | XP_005262915.1 | 47 - 48 | 7724 | 92 | |
XM_011531757.2 | XP_011530059.1 | 46 - 47 | 7635 | 92 | |
XM_011531759.2 | XP_011530061.1 | 45 - 46 | 7597 | 92 | |
XM_011531760.2 | XP_011530062.1 | 45 - 46 | 7535 | 92 | |
XM_011531761.2 | XP_011530063.1 | 46 - 47 | 7590 | 92 | |
XM_011531763.2 | XP_011530065.1 | 44 - 45 | 7481 | 92 | |
XM_011531764.2 | XP_011530066.1 | 45 - 46 | 7551 | 92 | |
XM_017007906.1 | XP_016863395.1 | 48 - 49 | 7852 | 92 | |
XM_017007907.1 | XP_016863396.1 | 48 - 49 | 7849 | 92 | |
XM_017007908.1 | XP_016863397.1 | 47 - 48 | 7781 | 92 | |
XM_017007909.1 | XP_016863398.1 | 33 - 34 | 5477 | 92 | |
GenBank mRNA | AB023210.2 | - | 10 - 11 | 1366 | 92 |
AF538685.1 | - | 44 - 45 | 7575 | 92 | |
BC150246.1 | - | 10 - 11 | 1387 | 92 | |
BX648077.1 | - | 2 - 3 | 248 | 92 | |
BX648153.1 | - | 5 - 6 | 576 | 92 |