The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 60 - 61 | 9613 | 65 |
XM_005262858.4 | XP_005262915.1 | 62 - 63 | 9654 | 65 | |
XM_011531757.2 | XP_011530059.1 | 61 - 62 | 9565 | 65 | |
XM_011531759.2 | XP_011530061.1 | 60 - 61 | 9527 | 65 | |
XM_011531760.2 | XP_011530062.1 | 60 - 61 | 9465 | 65 | |
XM_011531761.2 | XP_011530063.1 | 61 - 62 | 9520 | 65 | |
XM_011531762.2 | XP_011530064.1 | 60 - 61 | 9514 | 65 | |
XM_011531763.2 | XP_011530065.1 | 59 - 60 | 9411 | 65 | |
XM_011531764.2 | XP_011530066.1 | 60 - 61 | 9481 | 65 | |
XM_011531765.2 | XP_011530067.1 | 57 - 58 | 9216 | 65 | |
XM_017007906.1 | XP_016863395.1 | 63 - 64 | 9782 | 65 | |
XM_017007907.1 | XP_016863396.1 | 63 - 64 | 9779 | 65 | |
XM_017007908.1 | XP_016863397.1 | 62 - 63 | 9711 | 65 | |
XM_017007909.1 | XP_016863398.1 | 48 - 49 | 7407 | 65 | |
GenBank mRNA | AB023210.2 | - | 25 - 26 | 3296 | 65 |
AF538685.1 | - | 59 - 60 | 9505 | 65 | |
AK055806.1 | - | 11 - 12 | 1260 | 65 | |
BC065502.1 | - | 7 - 8 | 963 | 65 | |
BC150246.1 | - | 25 - 26 | 3317 | 65 | |
BX648077.1 | - | 17 - 18 | 2178 | 65 | |
BX648153.1 | - | 20 - 21 | 2506 | 65 |