The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161576.2 | NP_001155048.2 | 4 - 5 | 1218 | 138 |
NM_015589.5 | NP_056404.4 | 5 - 6 | 1482 | 138 | |
GenBank mRNA | AB028976.1 | - | 3 - 4 | 549 | 138 |
AK024652.1 | - | 5 - 6 | 1175 | 138 | |
AK025923.1 | - | 3 - 4 | 373 | 138 | |
BC121173.1 | - | 4 - 5 | 915 | 138 | |
BC121174.1 | - | 4 - 5 | 915 | 138 |