Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
Chromosome Location
Chr.1: 93179802 - 93279037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306076.1 NP_001293005.1 23 - 24 3361 141
NM_206886.4 NP_996769.3 23 - 24 3364 141
XM_011541361.2 XP_011539663.2 23 - 24 3573 141
XM_017001154.1 XP_016856643.1 23 - 24 3573 141
XM_017001155.1 XP_016856644.1 23 - 24 3570 141
XM_017001156.1 XP_016856645.1 23 - 24 3543 141
XM_017001157.1 XP_016856646.1 23 - 24 3525 141
XM_017001158.1 XP_016856647.1 23 - 24 3570 141
XM_017001159.1 XP_016856648.1 22 - 23 3441 141
XM_017001162.1 XP_016856651.1 23 - 24 3286 141
XM_017001163.1 XP_016856652.1 23 - 24 3840 141
XM_017001164.1 XP_016856653.1 23 - 24 3384 141
XM_017001165.1 XP_016856654.1 23 - 24 3573 141
XM_017001166.1 XP_016856655.1 21 - 22 3028 141
XM_017001167.1 XP_016856656.1 21 - 22 2963 141
XM_017001168.1 XP_016856657.1 19 - 20 2728 141
XM_017001170.1 XP_016856659.1 10 - 11 2957 141
XM_017001171.1 XP_016856660.1 10 - 11 1255 141
GenBank mRNA AK126045.1 - 17 - 18 2432 141

View Full Product Details