The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306076.1 | NP_001293005.1 | 27 - 28 | 4054 | 145 |
XM_011541361.2 | XP_011539663.2 | 27 - 28 | 4266 | 145 | |
XM_017001154.1 | XP_016856643.1 | 27 - 28 | 4266 | 145 | |
XM_017001155.1 | XP_016856644.1 | 27 - 28 | 4263 | 145 | |
XM_017001156.1 | XP_016856645.1 | 27 - 28 | 4236 | 145 | |
XM_017001157.1 | XP_016856646.1 | 27 - 28 | 4218 | 145 | |
XM_017001158.1 | XP_016856647.1 | 27 - 28 | 4263 | 145 | |
XM_017001159.1 | XP_016856648.1 | 26 - 27 | 4134 | 145 | |
XM_017001160.1 | XP_016856649.1 | 26 - 27 | 4122 | 145 | |
XM_017001161.1 | XP_016856650.1 | 26 - 27 | 4122 | 145 | |
XM_017001162.1 | XP_016856651.1 | 27 - 28 | 3979 | 145 | |
XM_017001163.1 | XP_016856652.1 | 27 - 28 | 4533 | 145 | |
XM_017001164.1 | XP_016856653.1 | 27 - 28 | 4077 | 145 | |
XM_017001166.1 | XP_016856655.1 | 25 - 26 | 3721 | 145 | |
XM_017001167.1 | XP_016856656.1 | 25 - 26 | 3656 | 145 | |
XM_017001168.1 | XP_016856657.1 | 23 - 24 | 3421 | 145 | |
XM_017001170.1 | XP_016856659.1 | 14 - 15 | 3650 | 145 | |
XM_017001171.1 | XP_016856660.1 | 14 - 15 | 1948 | 145 |