The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288629.1 | NP_001275558.1 | 7 - 8 | 1855 | 67 |
XM_017010365.1 | XP_016865854.1 | 7 - 8 | 1912 | 67 | |
GenBank mRNA | AB209269.1 | - | 7 - 8 | 1842 | 67 |
BC143857.1 | - | 7 - 8 | 1729 | 67 |