Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNC1H1
Entrez Gene ID
1778
Gene Name
dynein cytoplasmic 1 heavy chain 1
Gene Aliases
CMT2O, DHC1, DHC1a, DNCH1, DNCL, DNECL, DYHC, Dnchc1, HL-3, SMALED1, p22
Chromosome Location
Chr.14: 101964528 - 102050798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001376.4 NP_001367.2 24 - 25 5208 109
GenBank mRNA AB002323.3 - 24 - 25 5080 109
AB290157.1 - 24 - 25 5208 109
U53530.1 - 11 - 12 1658 109

View Full Product Details