The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001376.4 | NP_001367.2 | 26 - 27 | 5596 | 96 |
GenBank mRNA | AB002323.3 | - | 26 - 27 | 5468 | 96 |
AB290157.1 | - | 26 - 27 | 5596 | 96 | |
U53530.1 | - | 13 - 14 | 2046 | 96 |