The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001376.4 | NP_001367.2 | 3 - 4 | 682 | 114 |
GenBank mRNA | AB002323.3 | - | 3 - 4 | 554 | 114 |
AB290157.1 | - | 3 - 4 | 682 | 114 | |
AK299878.1 | - | 3 - 4 | 679 | 114 |