The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001376.4 | NP_001367.2 | 68 - 69 | 12559 | 58 |
GenBank mRNA | AB002323.3 | - | 68 - 69 | 12431 | 58 |
AB290157.1 | - | 68 - 69 | 12559 | 58 | |
AL833600.1 | - | 40 - 41 | 6557 | 58 | |
BC021297.1 | - | 11 - 12 | 1425 | 58 | |
BX648670.1 | - | 33 - 34 | 5161 | 58 |