The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009552.1 | NP_001009552.1 | 2 - 3 | 723 | 117 |
GenBank mRNA | AF444006.1 | - | 2 - 3 | 309 | 117 |
AK091875.1 | - | 3 - 4 | 487 | 117 | |
BC012022.1 | - | 2 - 3 | 334 | 117 | |
BT019526.1 | - | 2 - 3 | 308 | 117 | |
J03805.1 | - | 2 - 3 | 306 | 117 | |
X12656.1 | - | 2 - 3 | 329 | 117 |