The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166133.1 | NP_001159605.1 | 25 - 26 | 3591 | 93 |
NM_025074.6 | NP_079350.5 | 25 - 26 | 3591 | 93 | |
XM_006714316.2 | XP_006714379.1 | 25 - 26 | 3838 | 93 | |
GenBank mRNA | BC131820.1 | - | 25 - 26 | 3181 | 93 |