The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177531.4 | NP_803875.2 | 11 - 12 | 1028 | 148 |
XM_011517371.2 | XP_011515673.1 | 11 - 12 | 979 | 148 | |
XM_017013969.1 | XP_016869458.1 | 11 - 12 | 979 | 148 | |
XM_017013970.1 | XP_016869459.1 | 11 - 12 | 979 | 148 | |
XM_017013971.1 | XP_016869460.1 | 11 - 12 | 979 | 148 | |
XM_017013972.1 | XP_016869461.1 | 11 - 12 | 979 | 148 | |
GenBank mRNA | AY219181.1 | - | 11 - 12 | 1028 | 148 |