The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177531.4 | NP_803875.2 | 30 - 31 | 3734 | 85 |
XM_011517371.2 | XP_011515673.1 | 30 - 31 | 3685 | 85 | |
XM_017013969.1 | XP_016869458.1 | 30 - 31 | 3685 | 85 | |
XM_017013970.1 | XP_016869459.1 | 29 - 30 | 3571 | 85 | |
XM_017013971.1 | XP_016869460.1 | 30 - 31 | 3685 | 85 | |
XM_017013972.1 | XP_016869461.1 | 28 - 29 | 3382 | 85 | |
XM_017013973.1 | XP_016869462.1 | 6 - 7 | 741 | 85 | |
GenBank mRNA | AY219181.1 | - | 30 - 31 | 3734 | 85 |