Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKHD1L1
Entrez Gene ID
93035
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
Gene Aliases
PKHDL1
Chromosome Location
Chr.8: 109362477 - 109536809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177531.4 NP_803875.2 52 - 53 9020 78
XM_011517371.2 XP_011515673.1 52 - 53 8971 78
XM_017013969.1 XP_016869458.1 52 - 53 8971 78
XM_017013970.1 XP_016869459.1 51 - 52 8857 78
XM_017013971.1 XP_016869460.1 52 - 53 8809 78
XM_017013972.1 XP_016869461.1 50 - 51 8668 78
XM_017013973.1 XP_016869462.1 28 - 29 6027 78
XM_017013974.1 XP_016869463.1 23 - 24 5527 78
GenBank mRNA AY219181.1 - 52 - 53 9020 78

View Full Product Details