The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199378.1 | NP_001186307.1 | 17 - 18 | 2401 | 81 |
NM_001199379.1 | NP_001186308.1 | 17 - 18 | 2398 | 81 | |
NM_004193.2 | NP_004184.1 | 17 - 18 | 2398 | 81 | |
XM_005270261.1 | XP_005270318.1 | 17 - 18 | 2401 | 81 | |
XM_006718047.1 | XP_006718110.1 | 18 - 19 | 2500 | 81 | |
XM_006718048.1 | XP_006718111.1 | 18 - 19 | 2497 | 81 | |
XM_006718049.1 | XP_006718112.1 | 18 - 19 | 2500 | 81 | |
XM_006718050.1 | XP_006718113.1 | 18 - 19 | 2464 | 81 | |
XM_011540312.1 | XP_011538614.1 | 17 - 18 | 2359 | 81 | |
XM_011540313.1 | XP_011538615.1 | 18 - 19 | 2500 | 81 | |
XM_011540314.2 | XP_011538616.1 | 6 - 7 | 878 | 81 | |
XM_017016861.1 | XP_016872350.1 | 18 - 19 | 2476 | 81 | |
XM_017016862.1 | XP_016872351.1 | 18 - 19 | 2464 | 81 | |
XM_017016863.1 | XP_016872352.1 | 17 - 18 | 2362 | 81 | |
XM_017016864.1 | XP_016872353.1 | 13 - 14 | 1729 | 81 | |
GenBank mRNA | AF068755.1 | - | 17 - 18 | 2344 | 81 |
AK025330.1 | - | 17 - 18 | 2367 | 81 | |
BC117681.1 | - | 17 - 18 | 2225 | 81 | |
BC117682.1 | - | 17 - 18 | 2225 | 81 | |
D87435.2 | - | 17 - 18 | 2328 | 81 |