The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199378.1 | NP_001186307.1 | 25 - 26 | 3515 | 67 |
NM_001199379.1 | NP_001186308.1 | 25 - 26 | 3512 | 67 | |
NM_004193.2 | NP_004184.1 | 25 - 26 | 3512 | 67 | |
XM_005270261.1 | XP_005270318.1 | 25 - 26 | 3515 | 67 | |
XM_006718047.1 | XP_006718110.1 | 26 - 27 | 3614 | 67 | |
XM_006718048.1 | XP_006718111.1 | 26 - 27 | 3611 | 67 | |
XM_006718049.1 | XP_006718112.1 | 26 - 27 | 3614 | 67 | |
XM_006718050.1 | XP_006718113.1 | 26 - 27 | 3578 | 67 | |
XM_011540312.1 | XP_011538614.1 | 25 - 26 | 3473 | 67 | |
XM_011540313.1 | XP_011538615.1 | 26 - 27 | 3614 | 67 | |
XM_011540314.2 | XP_011538616.1 | 14 - 15 | 1992 | 67 | |
XM_017016861.1 | XP_016872350.1 | 26 - 27 | 3590 | 67 | |
XM_017016862.1 | XP_016872351.1 | 26 - 27 | 3578 | 67 | |
XM_017016863.1 | XP_016872352.1 | 25 - 26 | 3476 | 67 | |
XM_017016864.1 | XP_016872353.1 | 21 - 22 | 2814 | 67 | |
GenBank mRNA | AF068755.1 | - | 25 - 26 | 3458 | 67 |
AK024916.1 | - | 5 - 6 | 625 | 67 | |
AK025330.1 | - | 25 - 26 | 3481 | 67 | |
BC117681.1 | - | 25 - 26 | 3339 | 67 | |
BC117682.1 | - | 25 - 26 | 3339 | 67 | |
D87435.2 | - | 25 - 26 | 3442 | 67 |