The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014978.2 | NP_055793.1 | 15 - 16 | 2357 | 98 |
XM_011539542.2 | XP_011537844.1 | 10 - 11 | 1108 | 98 | |
GenBank mRNA | AB028982.1 | - | 15 - 16 | 2357 | 98 |
AK303009.1 | - | 2 - 3 | 421 | 98 | |
BC117417.1 | - | 15 - 16 | 2173 | 98 |