The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014978.2 | NP_055793.1 | 20 - 21 | 3089 | 69 |
XM_011539542.2 | XP_011537844.1 | 15 - 16 | 1840 | 69 | |
GenBank mRNA | AB028982.1 | - | 20 - 21 | 3089 | 69 |
AK303009.1 | - | 8 - 9 | 1275 | 69 | |
BC117417.1 | - | 20 - 21 | 2905 | 69 |