The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170585.1 | NP_001164056.1 | 54 - 55 | 4009 | 95 |
NM_153021.4 | NP_694566.4 | 55 - 56 | 4042 | 95 | |
XM_011532581.2 | XP_011530883.1 | 57 - 58 | 4312 | 95 | |
XM_011532584.2 | XP_011530886.1 | 57 - 58 | 4349 | 95 | |
XM_011532589.2 | XP_011530891.1 | 57 - 58 | 4346 | 95 | |
XM_011532591.2 | XP_011530893.1 | 57 - 58 | 4348 | 95 | |
XM_011532593.2 | XP_011530895.1 | 53 - 54 | 3924 | 95 | |
XM_011532601.2 | XP_011530903.1 | 43 - 44 | 3337 | 95 | |
XM_011532602.2 | XP_011530904.1 | 42 - 43 | 3309 | 95 | |
XM_011532603.2 | XP_011530905.1 | 42 - 43 | 3386 | 95 | |
XM_011532609.2 | XP_011530911.1 | 40 - 41 | 3140 | 95 | |
XM_011532610.2 | XP_011530912.1 | 32 - 33 | 3326 | 95 | |
XM_017003432.1 | XP_016858921.1 | 56 - 57 | 4318 | 95 | |
XM_017003433.1 | XP_016858922.1 | 56 - 57 | 4284 | 95 | |
GenBank mRNA | BC042674.1 | - | 15 - 16 | 1088 | 95 |
BC150633.1 | - | 55 - 56 | 4042 | 95 | |
BC153864.1 | - | 54 - 55 | 3961 | 95 |