The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024688.2 | NP_001019859.1 | 14 - 15 | 2286 | 183 |
NM_002485.4 | NP_002476.2 | 13 - 14 | 2183 | 183 | |
XM_011517045.1 | XP_011515347.1 | 14 - 15 | 2116 | 183 | |
XM_017013460.1 | XP_016868949.1 | 15 - 16 | 2267 | 183 | |
XM_017013461.1 | XP_016868950.1 | 12 - 13 | 2006 | 183 | |
XM_017013462.1 | XP_016868951.1 | 13 - 14 | 2191 | 183 | |
GenBank mRNA | AF051334.1 | - | 13 - 14 | 2124 | 183 |
AF058696.2 | - | 13 - 14 | 2125 | 183 | |
AK223256.1 | - | 13 - 14 | 2127 | 183 | |
AK289848.1 | - | 13 - 14 | 2179 | 183 | |
AK312410.1 | - | 13 - 14 | 2185 | 183 | |
BC136802.1 | - | 13 - 14 | 2142 | 183 | |
BC136803.1 | - | 13 - 14 | 2142 | 183 | |
BC144413.1 | - | 13 - 14 | 2142 | 183 | |
BC146797.1 | - | 13 - 14 | 2135 | 183 | |
BX640816.1 | - | 14 - 15 | 2234 | 183 |