The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032294.2 | NP_115670.1 | 15 - 16 | 1590 | 110 |
NM_172206.1 | NP_757343.1 | 15 - 16 | 1536 | 110 | |
XM_005256817.1 | XP_005256874.1 | 16 - 17 | 1704 | 110 | |
XM_006721588.2 | XP_006721651.1 | 16 - 17 | 2264 | 110 | |
XM_011524033.1 | XP_011522335.1 | 14 - 15 | 1510 | 110 | |
XM_011524035.2 | XP_011522337.1 | 10 - 11 | 1031 | 110 | |
XM_017025219.1 | XP_016880708.1 | 15 - 16 | 1699 | 110 | |
XM_017025220.1 | XP_016880709.1 | 15 - 16 | 2150 | 110 | |
XM_017025221.1 | XP_016880710.1 | 8 - 9 | 772 | 110 | |
GenBank mRNA | AF425232.1 | - | 15 - 16 | 1543 | 110 |
AF425301.1 | - | 15 - 16 | 1536 | 110 | |
AL136576.1 | - | 15 - 16 | 1590 | 110 | |
BC043487.1 | - | 15 - 16 | 1566 | 110 |