The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005923.3 | NP_005914.1 | 17 - 18 | 2781 | 152 |
XM_011535839.2 | XP_011534141.2 | 18 - 19 | 3105 | 152 | |
XM_017010870.1 | XP_016866359.1 | 18 - 19 | 3104 | 152 | |
XM_017010871.1 | XP_016866360.1 | 18 - 19 | 3104 | 152 | |
XM_017010872.1 | XP_016866361.1 | 18 - 19 | 3103 | 152 | |
XM_017010873.1 | XP_016866362.1 | 18 - 19 | 3102 | 152 | |
XM_017010874.1 | XP_016866363.1 | 17 - 18 | 2996 | 152 | |
XM_017010875.1 | XP_016866364.1 | 17 - 18 | 2995 | 152 | |
XM_017010876.1 | XP_016866365.1 | 16 - 17 | 2112 | 152 | |
XM_017010877.1 | XP_016866366.1 | 16 - 17 | 2349 | 152 | |
GenBank mRNA | AB209093.1 | - | 17 - 18 | 2885 | 152 |
AK293953.1 | - | 17 - 18 | 2041 | 152 | |
AK294507.1 | - | 7 - 8 | 763 | 152 | |
BC054503.2 | - | 17 - 18 | 2728 | 152 | |
BC088829.1 | - | 17 - 18 | 2782 | 152 | |
D84476.2 | - | 17 - 18 | 2687 | 152 | |
U67156.1 | - | 17 - 18 | 2780 | 152 |