Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K5
Entrez Gene ID
4217
Gene Name
mitogen-activated protein kinase kinase kinase 5
Gene Aliases
ASK1, MAPKKK5, MEKK5
Chromosome Location
Chr.6: 136557046 - 136793098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005923.3 NP_005914.1 26 - 27 4127 97
XM_017010870.1 XP_016866359.1 27 - 28 4453 97
XM_017010871.1 XP_016866360.1 27 - 28 4450 97
XM_017010872.1 XP_016866361.1 27 - 28 4452 97
XM_017010873.1 XP_016866362.1 27 - 28 4448 97
XM_017010874.1 XP_016866363.1 26 - 27 4345 97
XM_017010875.1 XP_016866364.1 26 - 27 4341 97
XM_017010876.1 XP_016866365.1 25 - 26 3461 97
XM_017010877.1 XP_016866366.1 25 - 26 3698 97
GenBank mRNA AB209093.1 - 26 - 27 4234 97
AK293953.1 - 26 - 27 3390 97
AK294507.1 - 16 - 17 2109 97
BC054503.2 - 26 - 27 4074 97
BC088829.1 - 26 - 27 4128 97
D84476.2 - 26 - 27 4033 97
U67156.1 - 26 - 27 4126 97

View Full Product Details