The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033280.3 | NP_150596.1 | 5 - 6 | 616 | 148 |
XM_011526260.2 | XP_011524562.1 | 6 - 7 | 623 | 148 | |
XM_017026073.1 | XP_016881562.1 | 5 - 6 | 477 | 148 | |
GenBank mRNA | AF212233.1 | - | 5 - 6 | 581 | 148 |
BC009703.1 | - | 5 - 6 | 568 | 148 | |
BM760252.1 | - | 3 - 4 | 207 | 148 |