The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300960.1 | NP_001287889.1 | 11 - 12 | 1422 | 80 |
NM_001300963.1 | NP_001287892.1 | 13 - 14 | 1370 | 80 | |
NM_001300964.1 | NP_001287893.1 | 12 - 13 | 1324 | 80 | |
NM_015076.4 | NP_055891.1 | 12 - 13 | 1554 | 80 | |
XM_005266871.3 | XP_005266928.1 | 9 - 10 | 1137 | 80 | |
XM_011535630.2 | XP_011533932.1 | 12 - 13 | 1900 | 80 | |
XM_011535631.2 | XP_011533933.1 | 11 - 12 | 1772 | 80 | |
XM_017010587.1 | XP_016866076.1 | 13 - 14 | 1780 | 80 | |
XM_017010588.1 | XP_016866077.1 | 10 - 11 | 1246 | 80 | |
GenBank mRNA | AB028951.2 | - | 12 - 13 | 1371 | 80 |
AK292360.1 | - | 12 - 13 | 1462 | 80 | |
AK300567.1 | - | 9 - 10 | 1046 | 80 | |
AL122055.1 | - | 9 - 10 | 1009 | 80 | |
AY028424.1 | - | 12 - 13 | 1554 | 80 | |
BC024247.1 | - | 13 - 14 | 1375 | 80 | |
BC037289.1 | - | 12 - 13 | 1449 | 80 | |
JX028238.1 | - | 11 - 12 | 1242 | 80 |