The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300741.1 | NP_001287670.1 | 6 - 7 | 1326 | 57 |
NM_031438.3 | NP_113626.1 | 6 - 7 | 1380 | 57 | |
XM_005272095.1 | XP_005272152.1 | 6 - 7 | 1365 | 57 | |
XM_005272097.3 | XP_005272154.1 | 6 - 7 | 1326 | 57 | |
GenBank mRNA | AK098066.1 | - | 6 - 7 | 1349 | 57 |
AK304010.1 | - | 6 - 7 | 1314 | 57 | |
AL136592.1 | - | 6 - 7 | 1335 | 57 | |
BC026748.1 | - | 3 - 4 | 402 | 57 | |
BC041099.1 | - | 6 - 7 | 1364 | 57 |