The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153221.2 | NP_694953.2 | 6 - 7 | 1060 | 114 |
GenBank mRNA | AF542080.1 | - | 6 - 7 | 1028 | 114 |
AK127735.1 | - | 6 - 7 | 1095 | 114 | |
AK314213.1 | - | 6 - 7 | 1032 | 114 |