Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBX4
Entrez Gene ID
80714
Gene Name
PBX homeobox 4
Gene Aliases
-
Chromosome Location
Chr.19: 19561707 - 19618916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025245.2 NP_079521.1 6 - 7 1214 85
NR_038198.1 6 - 7 1113 85
XM_006722911.3 XP_006722974.1 5 - 6 795 85
XM_011528320.2 XP_011526622.1 7 - 8 1260 85
XM_011528322.2 XP_011526624.1 5 - 6 849 85
XM_017027326.1 XP_016882815.1 7 - 8 895 85
XM_017027327.1 XP_016882816.1 7 - 8 876 85
XM_017027329.1 XP_016882818.1 4 - 5 685 85
GenBank mRNA AJ300182.1 - 5 - 6 795 85
AK097427.1 - 6 - 7 951 85
BC141859.1 - 6 - 7 1214 85
BC143588.1 - 6 - 7 889 85
BC143589.1 - 6 - 7 900 85
CK300861.1 - 2 - 3 85

View Full Product Details