The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271602.1 | NP_001258531.1 | 7 - 8 | 888 | 76 |
NM_001318505.1 | NP_001305434.1 | 8 - 9 | 1143 | 76 | |
NM_014606.2 | NP_055421.1 | 8 - 9 | 1143 | 76 | |
XM_005263327.3 | XP_005263384.1 | 9 - 10 | 1270 | 76 | |
XM_005263329.3 | XP_005263386.1 | 8 - 9 | 1143 | 76 | |
XM_005263330.3 | XP_005263387.1 | 8 - 9 | 1143 | 76 | |
XM_005263331.1 | XP_005263388.1 | 8 - 9 | 1143 | 76 | |
XM_005263332.3 | XP_005263389.1 | 8 - 9 | 1143 | 76 | |
XM_017008806.1 | XP_016864295.1 | 9 - 10 | 1270 | 76 | |
XM_017008807.1 | XP_016864296.1 | 7 - 8 | 866 | 76 | |
GenBank mRNA | AK289990.1 | - | 8 - 9 | 1086 | 76 |
AK296379.1 | - | 7 - 8 | 864 | 76 | |
BC038960.1 | - | 8 - 9 | 1081 | 76 | |
D25215.1 | - | 8 - 9 | 1070 | 76 |