The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004688.2 | NP_004679.2 | 4 - 5 | 673 | 146 |
XM_005246941.2 | XP_005246998.1 | 4 - 5 | 536 | 146 | |
XM_017005247.1 | XP_016860736.1 | 4 - 5 | 746 | 146 | |
GenBank mRNA | AB451305.1 | - | 3 - 4 | 343 | 146 |
AB451436.1 | - | 3 - 4 | 343 | 146 | |
AK291548.1 | - | 4 - 5 | 661 | 146 | |
AK298754.1 | - | 4 - 5 | 661 | 146 | |
BC001268.1 | - | 4 - 5 | 393 | 146 | |
BC021987.2 | - | 3 - 4 | 349 | 146 | |
U32849.1 | - | 4 - 5 | 623 | 146 |