The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130782.2 | NP_570138.1 | 1 - 2 | 293 | 106 |
GenBank mRNA | AF076642.1 | - | 1 - 2 | 267 | 106 |
AF268036.1 | - | 1 - 2 | 279 | 106 | |
AK057114.1 | - | 1 - 2 | 286 | 106 | |
AK292015.1 | - | 1 - 2 | 285 | 106 | |
AK315377.1 | - | 1 - 2 | 286 | 106 | |
AV715708.1 | - | 1 - 2 | 299 | 106 | |
BC020632.1 | - | 1 - 2 | 285 | 106 |