The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002240.4 | NP_002231.1 | 2 - 3 | 614 | 71 |
GenBank mRNA | AJ001894.1 | - | 2 - 3 | 315 | 71 |
AK313997.1 | - | 2 - 3 | 87 | 71 | |
BC101547.1 | - | 2 - 3 | 87 | 71 | |
D87327.1 | - | 2 - 3 | 535 | 71 | |
DR000377.1 | - | 2 - 3 | 621 | 71 | |
U24660.1 | - | 2 - 3 | 244 | 71 |